Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001130442.2 | NP_001123914.1 |
NM_001318054.1 | NP_001304983.1 | |
NM_005343.3 | NP_005334.1 | |
NM_176795.4 | NP_789765.1 | |
GenBank mRNA | AB451336.1 | - |
AB451485.1 | - | |
AF493916.1 | - | |
AJ437024.1 | - | |
BC006499.2 | - | |
BC095471.1 | - | |
BM801600.1 | - | |
BM801632.1 | - | |
BM808879.1 | - | |
BQ574535.1 | - | |
BQ674260.1 | - | |
BT019421.1 | - | |
CR536579.1 | - | |
CR542271.1 | - | |
DN990077.1 | - | |
ENST00000397596.0 | - | |
KJ534867.1 | - | |
KJ534868.1 | - |