Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_004465.1 | NP_004456.1 |
XM_005248264.3 | XP_005248321.1 | |
GenBank mRNA | AB002097.1 | - |
AF411527.1 | - | |
AF508782.1 | - | |
AK023508.1 | - | |
AK315314.1 | - | |
BC069561.1 | - | |
BC105021.1 | - | |
BC105023.1 | - | |
BC143772.1 | - | |
CR541665.1 | - | |
CR541688.1 | - | |
ENST00000264664.0 | - | |
GQ351295.1 | - | |
U67918.1 | - |