Gene Transcripts

Gene Symbol
LHB
Entrez Gene ID
3972
Gene Name
luteinizing hormone beta polypeptide
Gene Aliases
CGB4, HH23, LSH-B, LSH-beta
Chromosome Location
Chr.19: 49015980 - 49017090 on Build GRCh38
Uni Gene
Hs.154704
Interrogated Sequence Translated Protein
RefSeq NM_000894.2 NP_000885.1
XM_011526975.1 XP_011525277.1
GenBank mRNA ENST00000221421.0 -

View Full Product Details