Gene Transcripts

Gene Symbol
CCL15
Entrez Gene ID
6359
Gene Name
C-C motif chemokine ligand 15
Gene Aliases
HCC-2, HMRP-2B, LKN-1, LKN1, MIP-1 delta, MIP-1D, MIP-5, MRP-2B, NCC-3, NCC3, SCYA15, SCYL3, SY15
Chromosome Location
Chr.17: 35997582 - 36002038 on Build GRCh38
Uni Gene
Hs.272493
Interrogated Sequence Translated Protein
RefSeq NM_032965.5 NP_116741.2
GenBank mRNA AF031587.1 -
AI244164.1 -
BX098368.1 -
BX497188.1 -
U58914.1 -

View Full Product Details