Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_000594.3 | NP_000585.2 |
GenBank mRNA | AB451492.1 | - |
AF043342.1 | - | |
AF098751.1 | - | |
AJ227911.1 | - | |
AJ251878.1 | - | |
AJ251879.1 | - | |
AK314960.1 | - | |
BC028148.1 | - | |
BI908079.1 | - | |
BP215875.1 | - | |
CA306559.1 | - | |
ENST00000449264.0 | - | |
FJ795028.1 | - | |
HQ201306.2 | - | |
M10988.1 | - | |
X01394.1 | - |