Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_005228.3 | NP_005219.2 |
NM_201282.1 | NP_958439.1 | |
NM_201283.1 | NP_958440.1 | |
NM_201284.1 | NP_958441.1 | |
GenBank mRNA | AB209442.1 | - |
AF125253.1 | - | |
AF277897.1 | - | |
AI217671.1 | - | |
AK127817.1 | - | |
AK225422.1 | - | |
AK290352.1 | - | |
AK294750.1 | - | |
AL598260.1 | - | |
AU137334.1 | - | |
AW163038.1 | - | |
AW295229.1 | - | |
AY550966.1 | - | |
AY698023.1 | - | |
AY698024.1 | - | |
BC070081.1 | - | |
BC094761.1 | - | |
BC118665.1 | - | |
BC128419.1 | - | |
CB160831.1 | - | |
DQ088980.1 | - | |
EF210210.1 | - | |
EF210211.1 | - | |
ENST00000275493.0 | - | |
GU228573.1 | - | |
GU228584.1 | - | |
GU255993.1 | - | |
HM437234.1 | - | |
HM437235.1 | - | |
HQ730875.1 | - | |
HQ912715.1 | - | |
HQ999997.1 | - | |
K03193.1 | - | |
U48722.1 | - | |
U51732.1 | - | |
U95089.1 | - | |
X00588.1 | - | |
X00663.1 | - |