Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_002176.3 | NP_002167.1 |
GenBank mRNA | AB451323.1 | - |
AB451452.1 | - | |
AB451491.1 | - | |
BC069314.1 | - | |
BC096150.3 | - | |
BC096151.2 | - | |
BC096152.2 | - | |
BC096153.2 | - | |
ENST00000380232.0 | - | |
M17373.2 | - | |
M25460.1 | - | |
M28622.1 | - | |
V00546.1 | - | |
V00547.1 | - |