Gene Transcripts

Gene Symbol
FLT3
Entrez Gene ID
2322
Gene Name
fms related tyrosine kinase 3
Gene Aliases
CD135, FLK-2, FLK2, STK1
Chromosome Location
Chr.13: 28003274 - 28100592 on Build GRCh38
Uni Gene
Hs.507590
Interrogated Sequence Translated Protein
RefSeq NM_004119.2 NP_004110.2
NR_130706.1
XM_011535015.2 XP_011533317.1
XM_011535017.2 XP_011533319.1
XM_011535018.2 XP_011533320.1
XM_017020486.1 XP_016875975.1
XM_017020487.1 XP_016875976.1
XM_017020488.1 XP_016875977.1
XM_017020489.1 XP_016875978.1
GenBank mRNA AW057705.1 -
AW772610.1 -
BC036028.1 -
BC126350.1 -
BC144039.1 -
BC144040.1 -
BI461248.1 -
ENST00000241453.0 -
L36162.1 -
U02687.1 -
Z26652.1 -

View Full Product Details