Gene Transcripts

Gene Symbol
BAX
Entrez Gene ID
581
Gene Name
BCL2 associated X, apoptosis regulator
Gene Aliases
BCL2L4
Chromosome Location
Chr.19: 48954825 - 48961798 on Build GRCh38
Uni Gene
Hs.624291
Interrogated Sequence Translated Protein
RefSeq NM_001291428.1 NP_001278357.1
NM_001291429.1 NP_001278358.1
NM_001291430.1 NP_001278359.1
NM_001291431.1 NP_001278360.1
NM_004324.3 NP_004315.1
NM_138761.3 NP_620116.1
NM_138763.3 NP_620118.1
NM_138764.4 NP_620119.2
NR_027882.1
XM_017027077.1 XP_016882566.1
GenBank mRNA AF007826.1 -
AF247393.1 -
AF250190.1 -
AI565203.1 -
AJ417988.1 -
AJ586909.1 -
AJ586910.1 -
AK291076.1 -
BC014175.2 -
BE396495.1 -
BM673184.1 -
BM706954.1 -
CN281003.1 -
ENST00000293288.0 -
HQ889830.1 -
JX524562.1 -
KF830846.1 -
KF830847.1 -
KF830848.1 -
L22473.1 -
L22474.1 -
L22475.1 -
T29044.1 -
U19599.1 -

View Full Product Details