Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_004322.3 | NP_004313.1 |
NM_032989.2 | NP_116784.1 | |
GenBank mRNA | AB451254.1 | - |
AB451378.1 | - | |
AF021792.1 | - | |
AF031523.1 | - | |
AK023420.1 | - | |
AK291863.1 | - | |
AK303051.1 | - | |
AK309150.1 | - | |
AW014069.1 | - | |
BC001901.2 | - | |
BC095431.1 | - | |
BE255791.1 | - | |
BG435829.1 | - | |
BG748336.1 | - | |
BT006678.1 | - | |
CR541935.1 | - | |
CR541959.1 | - | |
ENST00000394532.0 | - | |
U66879.1 | - |