Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_002253.2 | NP_002244.1 |
GenBank mRNA | AB209901.1 | - |
AF035121.1 | - | |
AF063658.1 | - | |
AK293668.1 | - | |
BC131822.1 | - | |
BP280621.1 | - | |
ENST00000263923.0 | - | |
EU826563.1 | - | |
FJ899739.1 | - | |
L04947.1 | - | |
X61656.1 | - |