Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_006343.2 | NP_006334.2 |
XM_005263565.4 | XP_005263622.1 | |
XM_005263568.4 | XP_005263625.1 | |
XM_011510490.2 | XP_011508792.1 | |
XM_017003164.1 | XP_016858653.1 | |
XM_017003165.1 | XP_016858654.1 | |
GenBank mRNA | AK026681.1 | - |
AK294196.1 | - | |
AK308695.1 | - | |
AK316584.1 | - | |
AL705756.1 | - | |
AW627665.1 | - | |
BC114917.1 | - | |
BC114918.1 | - | |
BM719997.1 | - | |
BX474860.1 | - | |
CA454601.1 | - | |
CN371299.1 | - | |
CN371300.1 | - | |
D16316.1 | - | |
U08023.1 | - |