Gene Transcripts

Gene Symbol
ZAP70
Entrez Gene ID
7535
Gene Name
zeta chain of T cell receptor associated protein kinase 70
Gene Aliases
ADMIO2, IMD48, SRK, STCD, STD, TZK, ZAP-70
Chromosome Location
Chr.2: 97712030 - 97744327 on Build GRCh38
Uni Gene
Hs.234569
Interrogated Sequence Translated Protein
RefSeq NM_001079.3 NP_001070.2
NM_207519.1 NP_997402.1
XM_017004867.1 XP_016860356.1
XM_017004868.1 XP_016860357.1
XM_017004869.1 XP_016860358.1
XM_017004870.1 XP_016860359.1
GenBank mRNA AB083211.1 -
AK303341.1 -
AK310628.1 -
AK310637.1 -
AL832287.1 -
BC039039.1 -
BC053878.1 -
ENST00000264972.0 -
L05148.1 -

View Full Product Details