Gene Transcripts

Gene Symbol
VAV1
Entrez Gene ID
7409
Gene Name
vav guanine nucleotide exchange factor 1
Gene Aliases
VAV
Chromosome Location
Chr.19: 6772668 - 6857366 on Build GRCh38
Uni Gene
Hs.116237
Interrogated Sequence Translated Protein
RefSeq NM_001258206.1 NP_001245135.1
NM_001258207.1 NP_001245136.1
NM_005428.3 NP_005419.2
XM_005259642.1 XP_005259699.1
GenBank mRNA AK225710.1 -
AK301128.1 -
AK313307.1 -
AW105491.1 -
BC013361.2 -
DC408645.1 -
ENST00000304076.0 -
X16316.1 -
X83931.1 -

View Full Product Details