Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_002264.3 | NP_002255.3 |
NR_026698.1 | ||
XM_005247437.3 | XP_005247494.1 | |
XM_005247439.3 | XP_005247496.1 | |
XM_011512795.2 | XP_011511097.1 | |
XM_011512797.2 | XP_011511099.1 | |
XM_017006363.1 | XP_016861852.1 | |
XM_017006364.1 | XP_016861853.1 | |
XM_017006365.1 | XP_016861854.1 | |
GenBank mRNA | AA689229.1 | - |
AF035311.1 | - | |
AI216375.1 | - | |
AK125626.1 | - | |
AK128280.1 | - | |
AK299030.1 | - | |
AK312713.1 | - | |
AL700464.1 | - | |
AL710311.1 | - | |
BC002374.2 | - | |
BC003009.1 | - | |
BC029937.1 | - | |
BC090864.1 | - | |
BG287593.1 | - | |
BT006959.1 | - | |
CA433573.1 | - | |
CR456743.1 | - | |
DA308391.1 | - | |
DN991078.1 | - | |
ENST00000344337.0 | - | |
S75295.1 | - | |
U20620.1 | - |