Gene Transcripts

Gene Symbol
MFHAS1
Entrez Gene ID
9258
Gene Name
malignant fibrous histiocytoma amplified sequence 1
Gene Aliases
LRRC65, MASL1
Chromosome Location
Chr.8: 8783354 - 8893621 on Build GRCh38
Uni Gene
Hs.379414
Interrogated Sequence Translated Protein
RefSeq NM_004225.2 NP_004216.2
XM_011543852.2 XP_011542154.1
GenBank mRNA AB016816.1 -
AK027007.1 -
AK096504.1 -
BC014226.2 -
ENST00000276282.0 -

View Full Product Details