Gene Transcripts

Gene Symbol
TNF
Entrez Gene ID
7124
Gene Name
tumor necrosis factor
Gene Aliases
DIF, TNF-alpha, TNFA, TNFSF2, TNLG1F
Chromosome Location
Chr.6: 31575567 - 31578336 on Build GRCh38
Uni Gene
Hs.241570
Interrogated Sequence Translated Protein
RefSeq NM_000594.3 NP_000585.2
GenBank mRNA AB451492.1 -
AF043342.1 -
AF098751.1 -
AJ227911.1 -
AJ251878.1 -
AJ251879.1 -
AK314960.1 -
BC028148.1 -
BI908079.1 -
BP215875.1 -
CA306559.1 -
ENST00000449264.0 -
FJ795028.1 -
HQ201306.2 -
M10988.1 -
X01394.1 -

View Full Product Details