Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_002745.4 | NP_002736.3 |
NM_138957.3 | NP_620407.1 | |
GenBank mRNA | AI004470.1 | - |
AK055080.1 | - | |
AK057143.1 | - | |
AK090869.1 | - | |
AK295188.1 | - | |
AK307615.1 | - | |
BC017832.1 | - | |
BC099905.1 | - | |
BQ774391.1 | - | |
DQ399292.1 | - | |
EF565108.1 | - | |
ENST00000215832.0 | - | |
Z11694.1 | - | |
Z11695.1 | - |