Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001206729.1 | NP_001193658.1 |
NM_005465.4 | NP_005456.1 | |
NM_181690.2 | NP_859029.1 | |
XM_005272994.4 | XP_005273051.1 | |
XM_005272995.2 | XP_005273052.1 | |
XM_006711726.3 | XP_006711789.1 | |
XM_011544012.2 | XP_011542314.1 | |
XM_011544013.2 | XP_011542315.1 | |
XM_011544014.2 | XP_011542316.1 | |
XM_016999985.1 | XP_016855474.1 | |
GenBank mRNA | AF085234.1 | - |
AF124141.1 | - | |
AF135794.1 | - | |
AJ245709.1 | - | |
AK055109.1 | - | |
AK308052.1 | - | |
AL117525.1 | - | |
AW960221.1 | - | |
AY005799.1 | - | |
BC019085.2 | - | |
BC020479.1 | - | |
BC121154.2 | - | |
BG702262.1 | - | |
ENST00000366539.0 | - | |
U79271.1 | - |