Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_002546.3 | NP_002537.3 |
GenBank mRNA | AB002146.1 | - |
AF134187.1 | - | |
AK223155.1 | - | |
AK308524.1 | - | |
AK313710.1 | - | |
BC011023.1 | - | |
BC030155.2 | - | |
BM748946.1 | - | |
BQ000897.1 | - | |
CK903484.1 | - | |
DA477521.1 | - | |
ENST00000297350.0 | - | |
U94332.1 | - |