Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_005585.4 | NP_005576.3 |
NR_027654.1 | ||
XM_011521561.2 | XP_011519863.1 | |
GenBank mRNA | AF035528.1 | - |
AF037469.1 | - | |
AF043640.1 | - | |
AM909653.1 | - | |
BC012986.2 | - | |
BC029288.1 | - | |
BC052569.1 | - | |
BU154759.1 | - | |
CA406301.1 | - | |
CB055216.1 | - | |
CN314453.1 | - | |
CN314455.1 | - | |
ENST00000288840.0 | - | |
U59914.1 | - |