Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001256090.1 | NP_001243019.1 |
NM_001256091.1 | NP_001243020.1 | |
NM_001256092.1 | NP_001243021.1 | |
NM_001256093.1 | NP_001243022.1 | |
NM_001256094.1 | NP_001243023.1 | |
NM_001880.3 | NP_001871.2 | |
NR_045768.1 | ||
NR_045769.1 | ||
NR_045770.1 | ||
NR_045771.1 | ||
NR_045772.1 | ||
NR_045773.1 | ||
NR_045774.1 | ||
GenBank mRNA | AF283776.2 | - |
AI200584.1 | - | |
AK128731.1 | - | |
AL050192.1 | - | |
AY029364.1 | - | |
BC026175.1 | - | |
BC107698.1 | - | |
BC130335.1 | - | |
BC130337.1 | - | |
BX648469.1 | - | |
DA995464.1 | - | |
DQ003036.1 | - | |
DQ003037.1 | - | |
DQ003038.1 | - | |
DQ003039.1 | - | |
DQ003040.1 | - | |
DQ003041.1 | - | |
DQ003042.1 | - | |
DQ003043.1 | - | |
DQ003044.1 | - | |
DQ003045.1 | - | |
DQ003046.1 | - | |
DQ003047.1 | - | |
DQ003048.1 | - | |
DQ003049.1 | - | |
ENST00000264110.0 | - | |
M31630.1 | - | |
X15875.1 | - |