Gene Transcripts

Gene Symbol
MERTK
Entrez Gene ID
10461
Gene Name
MER proto-oncogene, tyrosine kinase
Gene Aliases
MER, RP38, Tyro12, c-Eyk, c-mer
Chromosome Location
Chr.2: 111898479 - 112039946 on Build GRCh38
Uni Gene
Hs.306178
Interrogated Sequence Translated Protein
RefSeq NM_006343.2 NP_006334.2
XM_005263565.4 XP_005263622.1
XM_005263568.4 XP_005263625.1
XM_011510490.2 XP_011508792.1
XM_017003164.1 XP_016858653.1
XM_017003165.1 XP_016858654.1
GenBank mRNA AK026681.1 -
AK294196.1 -
AK308695.1 -
AK316584.1 -
AL705756.1 -
AW627665.1 -
BC114917.1 -
BC114918.1 -
BM719997.1 -
BX474860.1 -
CA454601.1 -
CN371299.1 -
CN371300.1 -
D16316.1 -
U08023.1 -

View Full Product Details