Gene Transcripts

Gene Symbol
CCDC13
Entrez Gene ID
152206
Gene Name
coiled-coil domain containing 13
Gene Aliases
-
Chromosome Location
Chr.3: 42704118 - 42773253 on Build GRCh38
Uni Gene
Hs.604182
Interrogated Sequence Translated Protein
RefSeq NM_144719.3 NP_653320.3
XM_005264898.3 XP_005264955.1
XM_005264899.4 XP_005264956.1
XM_011533418.2 XP_011531720.1
XM_011533419.2 XP_011531721.1
XM_011533420.2 XP_011531722.1
XM_011533421.2 XP_011531723.1
XM_017005781.1 XP_016861270.1
GenBank mRNA AK058196.1 -
AK302859.1 -
BC036050.1 -
BM664398.1 -
ENST00000310232.0 -

View Full Product Details