Gene Transcripts

Gene Symbol
Kdr
Entrez Gene ID
16542
Gene Name
kinase insert domain protein receptor
Gene Aliases
6130401C07, Flk-1, Flk1, Krd-1, Ly73, VEGFR-2, VEGFR2, sVEGFR-2
Chromosome Location
Chr.5: 75932827 - 75979074 on Build GRCm38
Uni Gene
Mm.285
Interrogated Sequence Translated Protein
RefSeq NM_010612.2 NP_034742.2
GenBank mRNA AK031739.1 -
AK054510.1 -
AK141938.1 -
BC020530.1 -
EU884114.1 -
S53103.1 -
X59397.1 -
X70842.1 -

View Full Product Details