Gene Transcripts

Gene Symbol
Smad6
Entrez Gene ID
17130
Gene Name
SMAD family member 6
Gene Aliases
Madh6, b2b390Clo
Chromosome Location
Chr.9: 63953076 - 64022085 on Build GRCm38
Uni Gene
Mm.325757
Interrogated Sequence Translated Protein
RefSeq NM_008542.3 NP_032568.3
GenBank mRNA AB221689.1 -
AF010133.1 -
AI415735.1 -
AK004671.1 -
AK046602.1 -
BC047280.1 -

View Full Product Details