Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001025093.2 | NP_001020264.1 |
NM_001284369.1 | NP_001271298.1 | |
NM_001284370.1 | NP_001271299.1 | |
NM_001284371.1 | NP_001271300.1 | |
NM_001284372.1 | NP_001271301.1 | |
NM_001284373.1 | NP_001271302.1 | |
NM_001284374.1 | NP_001271303.1 | |
NM_001284376.1 | NP_001271305.1 | |
NM_009715.3 | NP_033845.1 | |
GenBank mRNA | AF483482.1 | - |
AF483483.1 | - | |
AK035438.1 | - | |
AK041145.1 | - | |
AK046184.1 | - | |
AK047405.1 | - | |
AK051591.1 | - | |
AK087424.1 | - | |
AK139515.1 | - | |
AK139844.1 | - | |
AK140855.1 | - | |
AK144977.1 | - | |
AK145887.1 | - | |
AK146839.1 | - | |
AK218615.1 | - | |
AY902311.1 | - | |
BC042210.1 | - | |
BC079883.1 | - | |
BC082596.1 | - | |
BC085137.1 | - | |
BY474294.1 | - | |
M31629.1 | - | |
M77167.1 | - | |
S76657.1 | - | |
S76659.1 | - | |
U46026.1 | - |