Gene Transcripts

Gene Symbol
Crebbp
Entrez Gene ID
12914
Gene Name
CREB binding protein
Gene Aliases
AW558298, CBP, CBP/p300, KAT3A, p300/CBP
Chromosome Location
Chr.16: 4081334 - 4213454 on Build GRCm38
Uni Gene
Mm.132238
Interrogated Sequence Translated Protein
RefSeq NM_001025432.1 NP_001020603.1
GenBank mRNA AK048818.1 -
AK051071.1 -
AK136057.1 -
AK162593.1 -
BC010297.1 -
BC072594.1 -
BU705617.1 -
S66385.1 -

View Full Product Details