Gene Transcripts

Gene Symbol
Elk1
Entrez Gene ID
13712
Gene Name
ELK1, member of ETS oncogene family
Gene Aliases
Elk-1
Chromosome Location
Chr.X: 20933395 - 20950608 on Build GRCm38
Uni Gene
Mm.405823
Interrogated Sequence Translated Protein
RefSeq NM_007922.5 NP_031948.4
GenBank mRNA AK082260.1 -
AK132354.1 -
AK196053.1 -
AK216408.1 -
BC005706.1 -
BC054474.1 -
BY728635.1 -
CV561936.1 -
D26066.1 -
FN434115.1 -
X87257.1 -
Z36939.1 -

View Full Product Details