Gene Transcripts

Gene Symbol
Lmnb1
Entrez Gene ID
16906
Gene Name
lamin B1
Gene Aliases
-
Chromosome Location
Chr.18: 56707813 - 56753424 on Build GRCm38
Uni Gene
Mm.4105
Interrogated Sequence Translated Protein
RefSeq NM_010721.2 NP_034851.2
GenBank mRNA AK079511.1 -
AK089110.1 -
AK132252.1 -
AK180884.1 -
BC052729.1 -
BC058392.1 -
M35153.1 -
X16705.1 -

View Full Product Details