Gene Transcripts

Gene Symbol
Fgf10
Entrez Gene ID
14165
Gene Name
fibroblast growth factor 10
Gene Aliases
AEY17, BB213776, Fgf-10, Gsfaey17
Chromosome Location
Chr.13: 118714699 - 118792573 on Build GRCm38
Uni Gene
Mm.317323
Interrogated Sequence Translated Protein
RefSeq NM_008002.4 NP_032028.1
GenBank mRNA AB064594.1 -
AK034742.1 -
AK045267.1 -
AK085244.1 -
AK141633.1 -
BB487015.2 -
BC048229.1 -
D89080.1 -
U94517.1 -

View Full Product Details