Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001163815.1 | NP_001157287.1 |
NM_001163816.1 | NP_001157288.1 | |
NM_011691.4 | NP_035821.3 | |
GenBank mRNA | AK088586.1 | - |
AK132886.1 | - | |
AK146347.1 | - | |
AK150398.1 | - | |
AK151183.1 | - | |
AK151214.1 | - | |
AK151367.1 | - | |
AK151373.1 | - | |
AK151831.1 | - | |
AK152073.1 | - | |
AK152425.1 | - | |
AK152555.1 | - | |
AK153052.1 | - | |
AK153265.1 | - | |
AK154183.1 | - | |
AK159326.1 | - | |
AK169478.1 | - | |
AK171873.1 | - | |
BC020487.1 | - | |
BY139350.1 | - | |
BY496946.1 | - | |
CT010321.1 | - | |
D83266.1 | - | |
M59833.1 | - | |
X64361.1 | - |