Gene Transcripts

Gene Symbol
Akt3
Entrez Gene ID
23797
Gene Name
thymoma viral proto-oncogene 3
Gene Aliases
AI851531, D930002M15Rik, Nmf350
Chromosome Location
Chr.1: 177020076 - 177258289 on Build GRCm38
Uni Gene
Mm.235194
Interrogated Sequence Translated Protein
RefSeq NM_011785.3 NP_035915.3
GenBank mRNA AF124142.1 -
AK052953.1 -
AK148914.1 -
BC066861.1 -
CK390299.1 -

View Full Product Details