Gene Transcripts

Gene Symbol
Pik3c2b
Entrez Gene ID
240752
Gene Name
phosphoinositide-3-kinase, class 2, beta polypeptide
Gene Aliases
C330011J12Rik, Gm199, PI3K-C2beta, PI3KC2beta
Chromosome Location
Chr.1: 133045672 - 133108688 on Build GRCm38
Uni Gene
Mm.387308
Interrogated Sequence Translated Protein
RefSeq NM_001099276.2 NP_001092746.2
GenBank mRNA AK049181.1 -
AK165803.1 -
BC132141.1 -

View Full Product Details