Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_028384.1 | NP_082660.1 |
GenBank mRNA | AK013632.1 | - |
AK038868.1 | - | |
AK158659.1 | - | |
AW911821.1 | - | |
BB762565.1 | - | |
BB857029.1 | - | |
BC147354.1 | - | |
BG800529.1 | - | |
BM949737.1 | - | |
BY082969.1 | - | |
BY296983.1 | - |