Gene Transcripts

Gene Symbol
Tgfb1
Entrez Gene ID
21803
Gene Name
transforming growth factor, beta 1
Gene Aliases
TGF-beta1, TGFbeta1, Tgfb, Tgfb-1
Chromosome Location
Chr.7: 25687002 - 25704996 on Build GRCm38
Uni Gene
Mm.248380
Interrogated Sequence Translated Protein
RefSeq NM_011577.1 NP_035707.1
GenBank mRNA AJ009862.1 -
AK144163.1 -
BC013738.1 -
M13177.1 -

View Full Product Details