Gene Transcripts

Gene Symbol
Bax
Entrez Gene ID
12028
Gene Name
BCL2-associated X protein
Gene Aliases
-
Chromosome Location
Chr.7: 45461695 - 45466903 on Build GRCm38
Uni Gene
Mm.19904
Interrogated Sequence Translated Protein
RefSeq NM_007527.3 NP_031553.1
GenBank mRNA AK013298.1 -
AK149994.1 -
AK159233.1 -
AV355155.1 -
AY095934.1 -
BC018228.1 -
BC053380.1 -
BY308683.1 -
CJ078904.1 -
L22472.1 -

View Full Product Details