Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_007922.5 | NP_031948.4 |
GenBank mRNA | AK082260.1 | - |
AK132354.1 | - | |
AK196053.1 | - | |
AK216408.1 | - | |
BC005706.1 | - | |
BC054474.1 | - | |
BY728635.1 | - | |
CV561936.1 | - | |
D26066.1 | - | |
FN434115.1 | - | |
X87257.1 | - | |
Z36939.1 | - |