Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001130443.1 | NP_001123915.1 |
NM_001130444.1 | NP_001123916.1 | |
NM_008284.2 | NP_032310.2 | |
GenBank mRNA | AJ437023.1 | - |
AK183914.1 | - | |
AK218213.1 | - | |
AY373386.1 | - | |
BC011083.1 | - | |
BC050837.1 | - | |
BC061885.1 | - | |
BI731283.1 | - | |
BQ963436.1 | - | |
BU924724.1 | - | |
CK788792.1 | - |