Gene Transcripts

Gene Symbol
Mapk1
Entrez Gene ID
26413
Gene Name
mitogen-activated protein kinase 1
Gene Aliases
9030612K14Rik, AA407128, AU018647, C78273, ERK, Erk2, MAPK2, PRKM2, Prkm1, p41mapk, p42mapk
Chromosome Location
Chr.16: 16983382 - 17047453 on Build GRCm38
Uni Gene
Mm.196581
Interrogated Sequence Translated Protein
RefSeq NM_001038663.1 NP_001033752.1
NM_011949.3 NP_036079.1
GenBank mRNA AK013699.1 -
AK018549.1 -
AK035386.1 -
AK043621.1 -
AK048127.1 -
AK048159.1 -
AK087925.1 -
AK132241.1 -
AK148853.1 -
AK163692.1 -
AK193161.1 -
AK193200.1 -
AK205643.1 -
AK209313.1 -
AK210736.1 -
AK211534.1 -
AK216848.1 -
AK218813.1 -
BB648734.1 -
BC006708.1 -
BC058258.1 -
D10939.1 -
X58712.1 -

View Full Product Details