Gene Transcripts

Gene Symbol
Mfhas1
Entrez Gene ID
52065
Gene Name
malignant fibrous histiocytoma amplified sequence 1
Gene Aliases
2310066G09Rik, D8Ertd91e
Chromosome Location
Chr.8: 35587777 - 35679449 on Build GRCm38
Uni Gene
Mm.103246
Interrogated Sequence Translated Protein
RefSeq NM_001081279.1 NP_001074748.1
GenBank mRNA AK010068.1 -
AK035252.1 -
AK081646.1 -
AK132349.1 -
AK171731.1 -
AK171759.1 -
BC082308.1 -

View Full Product Details