Gene Transcripts

Gene Symbol
Ppap2b
Entrez Gene ID
67916
Gene Name
phosphatidic acid phosphatase type 2B
Gene Aliases
1110003O22Rik, 2610002D05Rik, AV025606, D4Bwg0538e, D4Bwg1535e, Lpp3, Ppab2b
Chromosome Location
Chr.4: 105157347 - 105232767 on Build GRCm38
Uni Gene
Mm.348326
Interrogated Sequence Translated Protein
RefSeq NM_080555.2 NP_542122.1
GenBank mRNA AK003390.1 -
AK011276.1 -
AK076183.1 -
AK159136.1 -
AK160052.1 -
AK160056.1 -
AK163427.1 -
AV018880.1 -
BC005558.1 -
BY129827.1 -

View Full Product Details