Gene Transcripts

Gene Symbol
Tnfrsf11b
Entrez Gene ID
18383
Gene Name
tumor necrosis factor receptor superfamily, member 11b (osteoprotegerin)
Gene Aliases
OCIF, Opg, TR1
Chromosome Location
Chr.15: 54250619 - 54278484 on Build GRCm38
Uni Gene
Mm.15383
Interrogated Sequence Translated Protein
RefSeq NM_008764.3 NP_032790.3
GenBank mRNA AB013898.1 -
AK140244.1 -
AK146105.1 -
BC049782.1 -
U94331.1 -

View Full Product Details