Gene Transcripts

Gene Symbol
Hras
Entrez Gene ID
15461
Gene Name
Harvey rat sarcoma virus oncogene
Gene Aliases
H-ras, Ha-ras, Harvey-ras, Hras-1, Hras1, Kras2, c-H-ras, c-Ha-ras, c-rasHa, ras
Chromosome Location
Chr.7: 141189934 - 141194004 on Build GRCm38
Uni Gene
Mm.334313
Interrogated Sequence Translated Protein
RefSeq NM_001130443.1 NP_001123915.1
NM_001130444.1 NP_001123916.1
NM_008284.2 NP_032310.2
GenBank mRNA AJ437023.1 -
AK183914.1 -
AK218213.1 -
AY373386.1 -
BC011083.1 -
BC050837.1 -
BC061885.1 -
BI731283.1 -
BQ963436.1 -
BU924724.1 -
CK788792.1 -

View Full Product Details