Gene Transcripts

Gene Symbol
Lhb
Entrez Gene ID
16866
Gene Name
luteinizing hormone beta
Gene Aliases
LH, LH-B, LSH-B, LSH-beta
Chromosome Location
Chr.7: 45418166 - 45421855 on Build GRCm38
Uni Gene
Mm.57061
Interrogated Sequence Translated Protein
RefSeq NM_008497.2 NP_032523.2
GenBank mRNA AV173269.2 -
BB618381.1 -
BM224915.2 -
Y10418.1 -

View Full Product Details