Gene Transcripts

Gene Symbol
Vav1
Entrez Gene ID
22324
Gene Name
vav 1 oncogene
Gene Aliases
Vav, vav-T
Chromosome Location
Chr.17: 57279079 - 57329236 on Build GRCm38
Uni Gene
Mm.248172
Interrogated Sequence Translated Protein
RefSeq NM_001163815.1 NP_001157287.1
NM_001163816.1 NP_001157288.1
NM_011691.4 NP_035821.3
GenBank mRNA AK088586.1 -
AK132886.1 -
AK146347.1 -
AK150398.1 -
AK151183.1 -
AK151214.1 -
AK151367.1 -
AK151373.1 -
AK151831.1 -
AK152073.1 -
AK152425.1 -
AK152555.1 -
AK153052.1 -
AK153265.1 -
AK154183.1 -
AK159326.1 -
AK169478.1 -
AK171873.1 -
BC020487.1 -
BY139350.1 -
BY496946.1 -
CT010321.1 -
D83266.1 -
M59833.1 -
X64361.1 -

View Full Product Details