Gene Transcripts

Gene Symbol
LOC433461
Entrez Gene ID
433461
Gene Name
uncharacterized LOC433461
Gene Aliases
-
Chromosome Location
-
Uni Gene
Mm.130401
Interrogated Sequence Translated Protein
GenBank mRNA AK051942.1 -

View Full Product Details