Gene Transcripts

Gene Symbol
CCL13
Entrez Gene ID
100033951
Gene Name
chemokine (C-C motif) ligand 13
Gene Aliases
MCP-4
Chromosome Location
Chr.11: 38804557 - 38805899 on Build EquCab3.0
Uni Gene
Eca.12653
Interrogated Sequence Translated Protein
RefSeq NM_001163887.1 NP_001157359.1
GenBank mRNA AJ251191.1 -

View Full Product Details