Gene Transcripts

Gene Symbol
TNFRSF10A
Entrez Gene ID
716826
Gene Name
TNF receptor superfamily member 10a
Gene Aliases
-
Chromosome Location
Chr.8: 23450474 - 23485616 on Build Mmul_10
Uni Gene
Mmu.27782
Interrogated Sequence Translated Protein
RefSeq XM_015144975.2 XP_015000461.2
GenBank mRNA JU495587.1 -
JV074496.1 -
JV664502.1 -

View Full Product Details