Gene Transcripts

Gene Symbol
PRKCG
Entrez Gene ID
5582
Gene Name
protein kinase C gamma
Gene Aliases
PKC-gamma, PKCC, PKCG, SCA14
Chromosome Location
Chr. 19: 53882184 - 53907647 on Build GRCh38
UniGene ID
Hs.631564
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316329.1 NP_001303258.1 5 823
NM_002739.4 NP_002730.1 5 823
GenBank mRNA AB209671.1 - 5 542
AF345987.1 - 5 707
AK296412.1 - 5 831
AK296434.1 - 5 810
AK302958.1 - 5 808
AK303741.1 - 3 230
AK312302.1 - 5 785
BC047876.1 - 5 698 5764695
M13977.1 - 5 707
Z15114.1 - 1 31

View Full Product Details